2-Bromo-6-fluoro-3-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: [2-Bromo-6-fluoro-3-(trifluoromethyl)phenyl]methanol

35267-1G 35267-500MG 35267-5G 35267-25G
76903-092EA 465.74 USD
76903-092 76903-094 76903-096 77635-680
2-Bromo-6-fluoro-3-(trifluoromethyl)benzyl alcohol ≥97%
2-Bromo-6-fluoro-3-(trifluoromethyl)benzyl alcohol
Formula: C₈H₅BrF₄O
MW: 273.022 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31580162
CAS Number: 2090690-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR