2-Fluoro-6-iodobenzoic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

35145-1G 35145-25G 35145-500MG 35145-5G
76903-078EA 265.23 USD
76903-078 76903-080 76903-082 76903-084
2-Fluoro-6-iodobenzoic acid ethyl ester ≥97%
2-Fluoro-6-iodobenzoic acid ethyl ester

Precaución: for Research Use Only

Formula: C₉H₈FIO₂
MW: 294.061 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12025053
CAS Number: 925215-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR