2,6-Dichloro-3-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

34965-1G 34965-25G 34965-500MG 34965-5G
76903-062EA 242.95 USD
76903-062 76903-064 76903-066 76903-068
2,6-Dichloro-3-phenylpyridine ≥97%
2,6-Dichloro-3-phenylpyridine
Formula: C₁₁H₇Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD00234689
CAS Number: 18700-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR