2-Bromo-6-fluoro-3-(trifluoromethyl)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

34892-1G 34892-500MG 34892-5G
76903-048EA 468.92 USD
76903-048 76903-050 76903-052
2-Bromo-6-fluoro-3-(trifluoromethyl)benzoic acid ≥97% research grade
2-Bromo-6-fluoro-3-(trifluoromethyl)benzoic acid
Formula: C₈H₃BrF₄O₂
MW: 287.006 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30497608
CAS Number: 2059937-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR