2-Bromo-4-butoxy-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

34762-25G 34762-5G
76902-986EA 346.92 USD
76902-986 76902-988
2-Bromo-4-butoxy-1-methylbenzene ≥97%
2-Bromo-4-butoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707002
CAS Number: 1856706-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR