2-Fluoro-4,5-dimethoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

34754-1G 34754-25G 34754-500MG 34754-5G
76902-972EA 439.22 USD
76902-972 76902-974 76902-976 76902-978
2-Fluoro-4,5-dimethoxyphenylboronic acid pinacol ester ≥97%
2-Fluoro-4,5-dimethoxyphenylboronic acid pinacol ester
Formula: C₁₄H₂₀BFO₄
MW: 282.116 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026083
CAS Number: 1150271-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR