2-Ethoxy-5-fluorobenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

34743-1G 34743-25G 34743-500MG 34743-5G
76902-956EA 173.99 USD
76902-956 76902-958 76902-960 76902-962
2-Ethoxy-5-fluorobenzaldehyde ≥97% research grade
2-Ethoxy-5-fluorobenzaldehyde
Formula: C₉H₉FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06247346
CAS Number: 5710-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR