3-(2-Bromophenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

34740-1G 34740-25G 34740-500MG 34740-5G
76902-942EA 350.1 USD
76902-942 76902-944 76902-946 76902-948
3-(2-Bromophenyl)pyridine ≥97%
3-(2-Bromophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD18415514
CAS Number: 847264-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR