2-Bromo-6-chloro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

34619-1G 34619-25G 34619-500MG 34619-5G
76902-914EA 142.17 USD
76902-914 76902-916 76902-918 76902-920
2-Bromo-6-chloro-3-methylbenzoic acid ≥97%
2-Bromo-6-chloro-3-methylbenzoic acid
Formula: C₈H₆BrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD31619515
CAS Number: 2091629-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR