2,5-Dibromobenzamide ≥97%

Supplier: AOBChem USA

34503-1G 34503-25G 34503-500MG 34503-5G
76902-890EA 194.15 USD
76902-890 76902-892 76902-894 76902-896
2,5-Dibromobenzamide ≥97%
2,5-Dibromobenzamide
Formula: C₇H₅Br₂NO
MDL Number: MFCD00672996
CAS Number: 170492-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR