2-Fluoro-3-phenylbenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

34476-1G 34476-25G 34476-500MG 34476-5G
76902-876EA 379.81 USD
76902-876 76902-878 76902-880 76902-882
2-Fluoro-3-phenylbenzaldehyde ≥97% research grade
2-Fluoro-3-phenylbenzaldehyde
Formula: C₁₃H₉FO
Storage Temperature: Ambient
MDL Number: MFCD26687658
CAS Number: 1508348-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR