2-Chloro-3-iodo-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

34359-25G 34359-5G
76902-854EA 453.01 USD
76902-854 76902-856
2-Chloro-3-iodo-4-methylpyridine ≥97%
2-Chloro-3-iodo-4-methylpyridine
Formula: C₆H₅ClIN
Storage Temperature: Ambient
MDL Number: MFCD11878566
CAS Number: 926922-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR