2-Formyl-4-phenylphenol ≥97%
Supplier: AOBChem USA
|
Warning
|
34131-1G
34131-25G
34131-500MG
34131-5G
76902-814EA
147.47
USD
76902-814
76902-816
76902-818
76902-820
2-Formyl-4-phenylphenol ≥97%
2-Formyl-4-phenylphenol
Formula:
C₁₃H₁₀O₂ MW: 198.217 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00579079 CAS Number: 1761-63-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...