2-Chloro-3-fluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

34052-1G 34052-500MG 34052-5G
76902-788EA 558.04 USD
76902-788 76902-790 76902-792
2-Chloro-3-fluoro-5-methylbenzaldehyde ≥97%
2-Chloro-3-fluoro-5-methylbenzaldehyde
Formula: C₈H₆ClFO
MW: 172.584 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28751750
CAS Number: 1805225-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR