2-Bromo-1-chloro-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

33826-1G 33826-25G 33826-500MG 33826-5G
76902-754EA 320.4 USD
76902-754 76902-756 76902-758 76902-760
2-Bromo-1-chloro-4-isopropoxybenzene ≥97%
2-Bromo-1-chloro-4-isopropoxybenzene
Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070762
CAS Number: 201849-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR