2,6-Dichloro-4-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

33798-1G 33798-500MG 33798-5G
76902-742EA 468.92 USD
76902-742 76902-744 76902-746
2,6-Dichloro-4-formylbenzoic acid ≥97%
2,6-Dichloro-4-formylbenzoic acid
Formula: C₈H₄Cl₂O₃
MW: 219.022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30829110
CAS Number: 1807998-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR