2-Fluoro-5-(trifluoromethyl)benzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-[2-Fluoro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(2-Fluoro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-5-(trifluoromethyl)phenylboronic acid pinacol ester, 2-Fluoro-5-(trifluoromethy)lphenylboronic acid pinacol ester, 1,3,2-Dioxaborolane, 2-[2-fluoro-5-(trifluoromethyl)phenyl]-4,4,5,5-tetramethyl-

33761-1G 33761-25G 33761-500MG 33761-5G
76902-726EA 130.5 USD
76902-726 76902-728 76902-730 76902-732
2-Fluoro-5-(trifluoromethyl)benzeneboronic acid pinacol ester ≥97%
2-Fluoro-5-(trifluoromethyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₄O₂
Storage Temperature: Ambient
MDL Number: MFCD12923173
CAS Number: 1192045-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR