2,4-Difluororesorcinol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Difluorobenzene-1,3-diol

33744-1G 33744-25G 33744-500MG 33744-5G
76902-704EA 144.29 USD
76902-704 76902-706 76902-708 76902-710
2,4-Difluororesorcinol ≥97%
2,4-Difluororesorcinol

Precaución: for Research Use Only

Formula: C₆H₄F₂O₂
Storage Temperature: Refrigerator
CAS Number: 195136-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR