2-Methylthiophene-3-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methylthiophene-3-carbaldehyde, 3-Formyl-2-methylthiophene

33719-1G 33719-25G 33719-500MG 33719-5G
76902-692EA 150.65 USD
76902-692 76902-694 76902-696 76902-698
2-Methylthiophene-3-carboxaldehyde ≥95%
2-Methylthiophene-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₆OS
Storage Temperature: Refrigerator
MDL Number: MFCD09864856
CAS Number: 84815-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR