2-Fluoro-4-methoxy-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

33618-1G 33618-25G 33618-500MG 33618-5G
76902-644EA 292.81 USD
76902-644 76902-646 76902-648 76902-650
2-Fluoro-4-methoxy-1,3-dimethylbenzene ≥97%
2-Fluoro-4-methoxy-1,3-dimethylbenzene
Formula: C₉H₁₁FO
Storage Temperature: Ambient
MDL Number: MFCD28794219
CAS Number: 1806418-49-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR