2,5-Dichloro-3-methoxybenzyl alcohol ≥97% research grade

Supplier: AOBChem USA

Warning

33591-1G 33591-25G 33591-500MG 33591-5G
76902-628EA 439.22 USD
76902-628 76902-630 76902-632 76902-634
2,5-Dichloro-3-methoxybenzyl alcohol ≥97% research grade
2,5-Dichloro-3-methoxybenzyl alcohol
Formula: C₈H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD31382557
CAS Number: 165377-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR