2,5-Dichloro-3-fluorobenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

33572-25G 33572-5G
76902-616EA 310.85 USD
76902-616 76902-618
2,5-Dichloro-3-fluorobenzoic acid ≥97% research grade
2,5-Dichloro-3-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD13250174
CAS Number: 501008-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR