2-Chloro-5-(trifluoromethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

33553-1G 33553-25G 33553-500MG 33553-5G
76902-600EA 153.84 USD
76902-600 76902-602 76902-604 76902-606
2-Chloro-5-(trifluoromethoxy)benzaldehyde ≥95%
2-Chloro-5-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄ClF₃O₂
MW: 224.564 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18393769
CAS Number: 468075-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR