2-Benzyloxy-4,5-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

33481-1G 33481-25G 33481-500MG 33481-5G
76902-570EA 254.62 USD
76902-570 76902-572 76902-574 76902-576
2-Benzyloxy-4,5-difluorophenylboronic acid ≥97%
2-Benzyloxy-4,5-difluorophenylboronic acid
Formula: C₁₃H₁₁BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD26960243
CAS Number: 2121512-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR