2-Fluoro-5-iodobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

33242-1G 33242-25G 33242-5G
76902-516EA 112.46 USD
76902-516 76902-518 76902-520
2-Fluoro-5-iodobenzyl alcohol ≥97%
2-Fluoro-5-iodobenzyl alcohol
Formula: C₇H₆FIO
Storage Temperature: Refrigerator
MDL Number: MFCD06202824
CAS Number: 438050-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR