2-Chloro-1-iodo-3-(trifluoromethyl)benzene ≥95% research grade

Supplier: AOBChem USA

Warning

33221-1G 33221-25G 33221-500MG 33221-5G
76902-504EA 290.69 USD
76902-504 76902-506 76902-508 76902-510
2-Chloro-1-iodo-3-(trifluoromethyl)benzene ≥95% research grade
2-Chloro-1-iodo-3-(trifluoromethyl)benzene
Formula: C₇H₃ClF₃I
Storage Temperature: Ambient
MDL Number: MFCD28753251
CAS Number: 1369918-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR