2-Chloro-6-methoxypyridine-4-boronic acid 95% research grade

Supplier: AOBChem USA

Warning

33199-1G 33199-500MG 33199-5G 33199-25G
76902-490EA 485.9 USD
76902-490 76902-492 76902-494 77655-120
2-Chloro-6-methoxypyridine-4-boronic acid 95% research grade
2-Chloro-6-methoxypyridine-4-boronic acid
Formula: C₆H₇BClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13188782
CAS Number: 2121512-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR