2,5-Difluoro-4-formylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Fifluoro-4-formylbenzoic acid

33172-1G 33172-500MG 33172-5G
76902-478EA 558.04 USD
76902-478 76902-480 76902-482
2,5-Difluoro-4-formylbenzoic acid ≥95%
2,5-Difluoro-4-formylbenzoic acid
Formula: C₈H₄F₂O₃
MW: 186.1124 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30497656
CAS Number: 1890953-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR