2-Bromo-5-chlorobenzoic acid ethyl ester ≥97%
Supplier: AOBChem USA
|
Warning
|
32894-1G
32894-25G
32894-500MG
32894-5G
76902-426EA
238.71
USD
76902-426
76902-428
76902-430
76902-432
2-Bromo-5-chlorobenzoic acid ethyl ester ≥97%
2-Bromo-5-chlorobenzoic acid ethyl ester
Formula:
C₉H₈BrClO₂ MW: 263.516 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD10566825 CAS Number: 690260-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...