2-Methoxy-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Methoxy-3-methylphenyl)methanol

32670-25G 32670-5G
76902-394EA 233.4 USD
76902-394 76902-396
2-Methoxy-3-methylbenzyl alcohol ≥97%
2-Methoxy-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12031989
CAS Number: 74090-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR