2-Chloro-5-fluoro-4-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

32561-1G 32561-500MG 32561-5G
76902-384EA 528.33 USD
76902-384 76902-386 76902-388
2-Chloro-5-fluoro-4-methoxypyridine ≥97%
2-Chloro-5-fluoro-4-methoxypyridine
Formula: C₆H₅ClFNO
MW: 161.561 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18257279
CAS Number: 1211515-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR