2-Ethoxy-5-methoxy-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32551-25G 32551-5G
76902-376EA 595.17 USD
76902-376 76902-378
2-Ethoxy-5-methoxy-benzaldehyde ≥97%
2-Ethoxy-5-methoxy-benzaldehyde
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02380373
CAS Number: 39206-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR