2,6-Dibromo-4-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32539-1G 32539-25G 32539-5G
76902-366EA 76.39 USD
76902-366 76902-368 76902-370
2,6-Dibromo-4-fluorobenzoic acid ≥97%
2,6-Dibromo-4-fluorobenzoic acid
Formula: C₇H₃Br₂FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00460559
CAS Number: 91590-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR