2-Chloro-5-fluoro-3-(trifluoromethyl)benzoic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32497-1G 32497-25G 32497-500MG 32497-5G
76902-350EA 248.26 USD
76902-350 76902-352 76902-354 76902-356
2-Chloro-5-fluoro-3-(trifluoromethyl)benzoic acid pinacol ester ≥97%
2-Chloro-5-fluoro-3-(trifluoromethyl)benzoic acid pinacol ester
Formula: C₁₃H₁₄BClF₄O₂
MW: 324.507 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30529815
CAS Number: 2121515-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR