2-(2-Bromopyridin-4-yl)acetic acid ≥97%

Supplier: AOBChem USA

Warning

32482-1G 32482-25G 32482-5G
76902-336EA 62.6 USD
76902-336 76902-338 76902-340
2-(2-Bromopyridin-4-yl)acetic acid ≥97%
2-(2-Bromopyridin-4-yl)acetic acid
Formula: C₇H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD07367935
CAS Number: 183483-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR