2-Fluoro-3,4-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32466-1G 32466-25G 32466-500MG 32466-5G
76902-320EA 221.73 USD
76902-320 76902-322 76902-324 76902-326
2-Fluoro-3,4-dimethoxybenzaldehyde ≥97%
2-Fluoro-3,4-dimethoxybenzaldehyde
Formula: C₉H₉FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03411462
CAS Number: 37686-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR