2-Bromo-3,4-dimethoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32438-25G 32438-5G
76902-312EA 569.71 USD
76902-312 76902-314
2-Bromo-3,4-dimethoxybenzyl alcohol ≥97%
2-Bromo-3,4-dimethoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD02093896
CAS Number: 54370-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR