2,4-Dimethoxy-5-methylbenzaldehyde 97%

Supplier: AOBChem USA

Warning

32418-1G 32418-500MG 32418-5G 32418-25G
76902-286EA 1154.26 USD
76902-286 76902-288 76902-290 77655-050
2,4-Dimethoxy-5-methylbenzaldehyde 97%
2,4-Dimethoxy-5-methylbenzaldehyde
Formula: C₁₀H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00033853
CAS Number: 7149-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR