2-(2-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-3-methoxyphenylboronic acid pinacol ester

32397-25G 32397-5G
76902-274EA 488.02 USD
76902-274 76902-276
2-(2-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane ≥97%
2-(2-Chloro-3-methoxyphenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₈BClO₃
MW: 268.544 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782041
CAS Number: 1151564-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR