2,6-Dimethoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32293-1G 32293-25G 32293-5G
76902-260EA 84.88 USD
76902-260 76902-262 76902-264
2,6-Dimethoxybenzyl alcohol ≥97%
2,6-Dimethoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00016862
CAS Number: 16700-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR