2-Bromo-6-fluoro-4-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
32211-1G
32211-25G
32211-5G
76902-208EA
75.33
USD
76902-208
76902-210
76902-212
2-Bromo-6-fluoro-4-methylbenzoic acid ≥97%
2-Bromo-6-fluoro-4-methylbenzoic acid
Formula:
C₈H₆BrFO₂ MW: 233.03 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD23712068 CAS Number: 1427327-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...