2,4-Difluoro-m-toluic acid ≥97%

Supplier: AOBChem USA

Warning

32195-1G 32195-25G 32195-5G
76902-194EA 73.21 USD
76902-194 76902-196 76902-198
2,4-Difluoro-m-toluic acid ≥97%
2,4-Difluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03265223
CAS Number: 112857-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR