2-(2,5-Dibromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,5-Dibromophenylboronic acid pinacol ester
31927-1G
31927-25G
31927-5G
76902-138EA
64.72
USD
76902-138
76902-140
76902-142
2-(2,5-Dibromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2,5-Dibromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₂H₁₅BBr₂O₂ MW: 361.865 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17215817 CAS Number: 1256781-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...