2-Ethoxypyridine-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Ethoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

31882-1G 31882-25G 31882-500MG 31882-5G
76902-092EA 292.81 USD
76902-092 76902-094 76902-096 76902-098
2-Ethoxypyridine-4-boronic acid pinacol ester ≥97%
2-Ethoxypyridine-4-boronic acid pinacol ester
Formula: C₁₃H₂₀BNO₃
MW: 249.114 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11617874
CAS Number: 957346-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR