2,4-Difluoro-5-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

31729-1G 31729-25G 31729-500MG 31729-5G
76902-068EA 290.69 USD
76902-068 76902-070 76902-072 76902-074
2,4-Difluoro-5-methoxybenzoic acid ≥95%
2,4-Difluoro-5-methoxybenzoic acid
Formula: C₈H₆F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD28401421
CAS Number: 1266114-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR