2-(2-Chloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-methoxyphenylboronic acid pinacol ester

31679-25G 31679-5G
76902-042EA 616.39 USD
76902-042 76902-044
2-(2-Chloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(2-Chloro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₈BClO₃
MW: 268.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414622
CAS Number: 1416722-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR