2-Bromo-4,6-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

31617-1G 31617-25G 31617-500MG 31617-5G
76902-030EA 296 USD
76902-030 76902-032 76902-034 76902-036
2-Bromo-4,6-difluorobenzyl alcohol ≥97%
2-Bromo-4,6-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.015 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28737914
CAS Number: 1807071-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR