(2-Chloro-4-(hydroxymethyl)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-(hydroxymethyl)phenylboronic acid

31439-1G 31439-25G 31439-500MG 31439-5G
76902-000EA 224.92 USD
76902-000 76902-002 76902-004 76902-006
(2-Chloro-4-(hydroxymethyl)phenyl)boronic acid ≥97% research grade
(2-Chloro-4-(hydroxymethyl)phenyl)boronic acid
Formula: C₇H₈BClO₃
MW: 186.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805966
CAS Number: 2828440-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR