2-Methyl-4-(trifluoromethyl)benzyl alcohol ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: [2-Methyl-4-(trifluoromethyl)phenyl]methanol

31361-1G 31361-25G 31361-500MG 31361-5G
76901-968EA 201.58 USD
76901-968 76901-970 76901-972 76901-974
2-Methyl-4-(trifluoromethyl)benzyl alcohol ≥95% research grade
2-Methyl-4-(trifluoromethyl)benzyl alcohol
Formula: C₉H₉F₃O
MW: 190.162 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18399874
CAS Number: 888739-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR