2-Chloro-6-iodobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

31330-1G 31330-25G 31330-500MG 31330-5G
76901-954EA 177.18 USD
76901-954 76901-956 76901-958 76901-960
2-Chloro-6-iodobenzaldehyde ≥95%
2-Chloro-6-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClIO
MW: 266.464 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18393320
CAS Number: 51738-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR